Learn More
Invitrogen™ Prostasin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120449
Description
Positive test controls include: Mouse kidney. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass membrane protein, extracellular space, extracellular space. Immunogen sequence: TCNCLYNIDA KPEEPHFVQE DMVCAGYVEG GKDACQGDSG GPLSCPVEGL WYLTGIVSWG DACGARNRPG VYTLASSYAS WIQSKVTELQ PRVVPQTQES QPDSNLCGSH LAFSSAPAQG LLRPILFLPL GLALGLLSPW LSEH.
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
Specifications
| Prostasin | |
| Polyclonal | |
| Unconjugated | |
| PRSS8 | |
| 2410039E18Rik; AI313909; C79772; CAP1; channel activating protease 1; channel-activating protease 1; fr; mCAP1; PROSTASIN; Prostasin heavy chain; Prostasin light chain; protease, serine 8; protease, serine 8 (prostasin); protease, serine, 8; protease, serine, 8 (prostasin); Prss8; Serine protease 8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192107, 5652, 76560 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.06 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q16651, Q9ES87, Q9ESD1 | |
| PRSS8 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human PRSS8 (NP_002764.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.