missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prostasin Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580945
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Prostasin/PRSS8.
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
Specifications
| Prostasin | |
| Polyclonal | |
| Unconjugated | |
| PRSS8 | |
| 2410039E18Rik; AI313909; C79772; CAP1; channel activating protease 1; channel-activating protease 1; fr; mCAP1; PROSTASIN; Prostasin heavy chain; Prostasin light chain; protease, serine 8; protease, serine 8 (prostasin); protease, serine, 8; protease, serine, 8 (prostasin); Prss8; Serine protease 8 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5652 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q16651 | |
| PRSS8 | |
| Recombinant Human PRSS8 protein (Ala30-Arg322). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction