missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay

Product Code. 11903622
Change view
Click to view available options
Quantity:
96 Wells/Kit
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11903622 96 Wells/Kit 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11903622 Supplier Thermo Scientific™ Supplier No. R2474096
  • ONLINE EXCLUSIVE PRICE
    £842.40
    £823.46 / 96 tests
    Save £18.94
    2% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Rapidly detect E. coli Shiga Toxin 1 and 2 from human fecal samples.

Rapidly detect Shiga Toxin-producing Escherichia coli in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ STEC (Shiga Toxin E. coli) Microplate Assay.

Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Shiga Toxin-producing Escherichia coli. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT STEC (Shiga Toxin E.coli) is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT STEC provides same-day toxin detection that is easy-to-use, flexible and provides the power to do more with less.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description STEC Microplate Assay
Detectable Analytes E. coli Shiga Toxin 1 and 2
Sample Type Human Fecal
Type E. coli Testing Kit
For Use With (Application) Enzyme Immunoassay Detection
Includes Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity 96 Wells/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.