missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ProSpecT™ Campylobacter Microplate Assay
Description
Rapidly detect Campylobacter specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Campylobacter Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Campylobacter. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
- Excellent clinical performance
- Cary Blair transport option which saves a dilution step
- Results within 100 minutes
- Room temperature incubation
- True colorless negatives
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| CE Marker | Yes |
| Detectable Analytes | Campylobacter Antigen |
| Type | Campylobacter Microplate Assay |
| Quantity | 96 Tests/Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?