Learn More
Thermo Scientific™ ProSpecT™ Adenovirus Microplate Assay
Product Code. 13276079
Change view
Click to view available options
Quantity:
96 Tests/Kit
Unit Size:
96 tests
Product Code. Quantity unitSize
13276079 96 Tests/Kit 96 tests
1 options
Product Code. 13276079 Supplier Thermo Scientific™ Supplier No. R240096
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Rapidly detect Adenovirus specific antigen from human fecal samples.
Rapidly detect Adenovirus specific antigen in fresh or preserved human fecal specimens using Thermo Scientific™ ProSpecT™ Adenovirus Microplate Assay.
Manage your resources, increase productivity and improve workflow efficiency with this direct, qualitative enzyme immmunoassay for the detection of Adenovirus. It combines excellent sensitivity/specificity, a common procedure with no indeterminate range, plus flexible specimen transport options.
  • Excellent clinical performance
  • Cary Blair transport option which saves a dilution step
  • Results within 100 minutes
  • Room temperature incubation
  • True colorless negatives
ProSpecT Adenovirus is part of the most comprehensive line of microplate assays available for enteric infectious disease testing. The ProSpecT family of tests have a common format, common procedures and common reagents, maximizing operational efficiency, productivity and throughput. ProSpecT Adenovirus provides same-day detection that is easy-to-use, flexible and provides the power to do more with less.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Description ProSpecT Adenovirus Microplate Assay
Includes Transfer pipettes, microplate strip holder and cover, microplate (8 wells/strip), enzyme conjugate, positive control, negative control, sample diluent, wash buffer, color substrate, stop solution and procedure card
Quantity 96 Tests/Kit