missing translation for 'onlineSavingsMsg'
Learn More

PROSC Antibody, DyLight 350, Novus Biologicals™

Product Code. 18407009 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407009 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407009 Supplier Novus Biologicals Supplier No. NBP299607UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PROSC Polyclonal antibody specifically detects PROSC in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROSC
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias FLJ11861, proline synthase co-transcribed bacterial homolog protein, proline synthetase co-transcribed (bacterial homolog), proline synthetase co-transcribed bacterial homolog protein, proline synthetase co-transcribed homolog (bacterial)
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human PROSC.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.