missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes

Product Code. 17264523 Shop All Contec Products
Change view
Click to view available options
Dimensions (L x W):
15 x 23 cm (6 x 9 in.)
22.86 x 22.86 cm (9 x 9 in.)
23 x 28 cm (9 x 11 in.)
27.9 x 23 cm (11 x 9 in.)
Packaging Quantity:
20 Pk.
24 Pk.
30 Pk.
40 Packs of 20 Wipes
50 Pk.
Saturation Solution:
70% IPA, 30% DI water
70% USP Grade IPA, 30% DI Water
Unit Size:
Pack of 1152
Pack of 1400
Pack of 1440
Pack of 2000
Pack of 24
Pack of 800
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Packaging Quantity Saturation Solution unitSize
17264523 15 x 23 cm (6 x 9 in.) 50 Pk. 70% USP Grade IPA, 30% DI Water Pack of 2000
17224513 22.86 x 22.86 cm (9 x 9 in.) 20 Pk. 70% IPA, 30% DI water Pack of 800
17274513 23 x 28 cm (9 x 11 in.) 30 Pk. 70% IPA, 30% DI water Pack of 1440
17224523 23 x 28 cm (9 x 11 in.) 24 Pk. 70% IPA, 30% DI water Pack of 1152
17244523 23 x 28 cm (9 x 11 in.) 50 Pk. 70% USP Grade IPA, 30% DI Water Pack of 1400
11917025 27.9 x 23 cm (11 x 9 in.) 24 Pk. 70% IPA, 30% DI water Pack of 24
16294072 22.86 x 22.86 cm (9 x 9 in.) 40 Packs of 20 Wipes 70% IPA, 30% DI water Pack of 800
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17264523 Supplier Contec™ Supplier No. PSCP0001BPR

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Wipe down isolators, cabinets, hoods, and other cleanroom equipment and tools. Contec™ PROSAT™ Sterile Presaturated Nonwoven Wipes are ideal for cleanup following BD RODAC™ plate sampling. These wipes are designed specifically for sterile environments.

  • Designed for pharmaceutical, biotech, and medical device applications
  • Choose spunlaced nonwoven 100% polyester wipes for improved cleanliness, economical hydroentangled polyester/cellulose wipers, or meltblown polypropylene wipes for improved durability
  • Conveniently presaturated with 70/30 blend of semi-conductor grade IPA and DI water to reduce VOC emissions and hazardous waste
  • All wipes are sterilized by gamma radiation to a 10-6 Sterility Assurance Level and validated per AAMI ISO 11137 (Method 1) guidelines
  • Certificate of Sterility and Certificate of Irradiation included
  • Recommended for use in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Individually lot numbered resealable pouches with tamper-evident labels ensure product integrity and cleanliness

Compliance

Validated sterile to a Sterility Assurance Level of 10-6 in accordance with AAMI/ISO 1137 Method 1 Guidelines.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Pre-saturated Wipe
Length (English) 6 in.
Length (Metric) 15 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Polyester Cellulose
Certifications/Compliance ISO Class 4
Dimensions (L x W) 15 x 23 cm (6 x 9 in.)
For Use With (Application) Ideal for Routine Cleaning and Wipe Down of Lab Tools, Instruments and Other Equipment and Designed for Sterile Environments
Packaging Quantity 50 Pk.
Saturation Solution 70% USP Grade IPA, 30% DI Water
Sterility Sterile
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.