missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ProSAPiP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24936-1-AP-20UL
This item is not returnable.
View return policy
Description
Prosapip1 is a binding partner of the postsynaptic density (PSD) protein ProSAP2/Shank3, a major scaffolding protein in excitatory brain synapses. Prosapip1 is thought to bind ProSAP2/Shank3 through a PDZ domain in ProSAP2/Shank3 and link it in turn to the Spine-associated Rap-Gap (SPAR). Prosapip1 shares significant homology with another PSD protein, PSD-Zip70, with both coding for a Fez1 domain that can be found in a total of four related protein, forming a novel family of scaffolding molecules termed ′Fezzins′. Recent studies show that Prosapip1 can be found in endocrine cells in which it is localized primarily in the hormone-storing compartment of these cells, suggesting that Prosapip1 may have other functions in non-neuronal cells.Specifications
| ProSAPiP1 | |
| Polyclonal | |
| Unconjugated | |
| LZTS3 | |
| KIAA0552, ProSAP interacting protein 1, ProSAPiP1, ProSAPiP1 protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 241638, 9762 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.43 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| A2AHG0, O60299 | |
| LZTS3 | |
| ProSAPiP1 Fusion Protein Ag20765 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction