missing translation for 'onlineSavingsMsg'
Learn More

Propyltriphenylphosphonium iodide, 99%, Thermo Scientific™

Product Code. 15219836 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15219836

Brand: Thermo Scientific Acros 453040250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14350-50-6
Molecular Formula C21H22IP
Molecular Weight (g/mol) 432.29
MDL Number MFCD00050238
InChI Key MPNQDJZRGAOBPW-UHFFFAOYSA-M
Synonym triphenyl propyl phosphonium iodide, propyltriphenylphosphonium iodide, triphenyl propyl phosphanium iodide, propyl triphenyl phosphonium iodide, n-propyl-triphenyl phosphonium iodide, phosphonium,triphenylpropyl-, iodide 1:1
PubChem CID 10982905
IUPAC Name triphenyl(propyl)phosphanium;iodide
SMILES [I-].CCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 206.0°C to 209.0°C
Assay Percent Range 99%
Quantity 25 g
Formula Weight 432.28
Percent Purity 99%
Chemical Name or Material Propyltriphenylphosphonium iodide
compliance-icons
Product Identifier
  • Propyltriphenylphosphonium iodide, 99%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.