missing translation for 'onlineSavingsMsg'
Learn More

Propylthiouracil, MedChemExpress

Product Code. 16492106
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492106 500 mg 500mg
16488246 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492106 Supplier Medchem Express Supplier No. HYB0346
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propylthiouracil(6-Propyl-2-thiouracil) is a thyroperoxidase and 5'-deiodinase inhibitor.

Propylthiouracil(6-Propyl-2-thiouracil) is a thyroperoxidase and 5'-deiodinase inhibitor.

Specifications

Chemical Name or Material Propylthiouracil
Physical Form Solid
CAS 51-52-5
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C7H10N2OS
Synonym 6-n-Propylthiouracil 6-Propyl-2-thiouracil PTU
Solubility Information DMSO : ≥ 100 mg/mL (587.44 mM) ∣H2O : 0.67 mg/mL (3.94 mM; Need ultrasonic)
SMILES O=C(C=C(CCC)N1)NC1=S
Molecular Weight (g/mol) 170.23
Formula Weight 170.23
Percent Purity 99.2%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.