missing translation for 'onlineSavingsMsg'
Learn More

Propylenediphosphonic acid, 98+%

Product Code. 11387238
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387238 2 g 2g
11397238 0.5 g 0.5g
11307248 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11387238 Supplier Thermo Scientific Alfa Aesar Supplier No. 030277.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4671-82-3 | C3H10O6P2 | 204.055 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4671-82-3
Molecular Formula C3H10O6P2
Molecular Weight (g/mol) 204.055
MDL Number MFCD00050066
InChI Key DZQCAOQMXPROIJ-UHFFFAOYSA-N
Synonym propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
PubChem CID 22023450
IUPAC Name 1-phosphonopropan-2-ylphosphonic acid
SMILES CC(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 178°C
Odor Odorless
Quantity 2 g
Formula Weight 204.06
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Propylenediphosphonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.