missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol, Multi-Compendial, U.S.P., J.T. Baker™

Product Code. p-5391330 Shop All J.T.Baker Products
Click to view available options
Quantity:
4 L
19 L
Packaging:
Narrow Mouth Amber Glass Bottle
Poly Pail
Unit Size:
19 L
4 L
This item is not returnable. View return policy

Product Code. 15508614

Brand: Avantor J.T.Baker 940303

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-55-6
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key DNIAPMSPPWPWGF-UHFFFAOYSA-N
Synonym 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost
PubChem CID 1030
ChEBI CHEBI:16997
IUPAC Name propane-1,2-diol
SMILES CC(CO)O

Specifications

Quantity 4 L
Formula Weight 76.1
Flash Point 99.00°C
Grade Multi-Compendial/USP
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.04kg
Chemical Name or Material Propylene Glycol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.