missing translation for 'onlineSavingsMsg'
Learn More

Propylene Glycol (1,2 Propanediol), ACS/USP/NF, Greenfield

Product Code. 30286166
Change view
Click to view available options
Packaging:
4x1 Gallon Case
4x4L Case
Unit Size:
15.14 L
16 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
30286166 4x1 Gallon Case 15.14 L
30286174 4x4L Case 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30286166 Supplier Pharmco Supplier No. 369000000CSGL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-55-6

Specifications

Grade ACS
Packaging 4x1 Gallon Case
Quantity 4x1 Gallon Case
Chemical Name or Material Propylene Glycol (1,2 Propanediol), ACS/USP/NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.