missing translation for 'onlineSavingsMsg'
Learn More

Propylene carbonate, 99%

Product Code. 15821637
Change view
Click to view available options
Quantity:
250 g
1000 g
2.5 kg
5 kg
Unit Size:
1000g
250g
5000g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15821637 2.5 kg -
11412997 250 g 250g
11402997 1000 g 1000g
11422997 5 kg 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15821637 Supplier Thermo Scientific Alfa Aesar Supplier No. A15552.A4
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-32-7 | C4H6O3 | 102.09 g/mol

Cobalt(II) acetate tetrahydrate is used to prepare complexes for examination of the properties of metals with unusual coordination geometries. It is also used as a catalyst for oxidation and esterification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Propylene carbonate is a polar, aprotic solvent used in paints and coatings and as a high-permittivity component of electrolytes in lithium batteries. It plays an important role in adhesives, paint strippers, cosmetics, plasticizer, hard surface cleaner, resin cleaner, fiberglass, polyester and polyurethane cleaner. Furthermore, it acts as a cleaner in polyurethane, a carburetor and is used in stereo lithography parts. In electrospray ionization mass spectrometry, it is doped into low surface tension solutions to enhance analyte charging.

Solubility
Miscible with water, alcohol, ether, acetone, benzene, chloroform and ethyl acetate.

Notes
Moisture sensitive. Hygroscopic. Incompatible with acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-32-7
Molecular Formula C4H6O3
Molecular Weight (g/mol) 102.09
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
PubChem CID 7924
IUPAC Name 4-methyl-1,3-dioxolan-2-one
SMILES CC1COC(=O)O1

Specifications

Melting Point °C to 49°C
Density 1.204
Boiling Point 240°C to 243°C
Flash Point 132°C
Assay Percent Range 99%
Refractive Index 1.421
Quantity 2.5 kg
Solubility Information Miscible with water,alcohol,ether,acetone,benzene,chloroform and ethyl acetate.
Formula Weight 102.09
Physical Form Liquid
Chemical Name or Material Propylene Carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Propylene carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Propylene carbonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.