missing translation for 'onlineSavingsMsg'
Learn More

Propyl pyrazole triol, MedChemExpress

Product Code. 16425857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16425857 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16425857 Supplier Medchem Express Supplier No. HY100689
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%).

Propyl pyrazole triol (PPT) is a selective estrogen receptor alpha (ERα) agonist. The relative binding affinity of Propyl pyrazole triol for ERα (ERα: 49%) around 410 times higher compared with estrogen receptor beta (ERβ: 0.12%).

Specifications

Chemical Name or Material Propyl pyrazole triol
CAS 263717-53-9
Color White
Purity Grade Notes Research
Molecular Formula C24H22N2O3
Quantity 5 mg
Synonym PPT
Solubility Information DMSO : ≥ 100 mg/mL (258.77 mM)
SMILES CCCC1=C(C2=CC=C(O)C=C2)N(C3=CC=C(O)C=C3)N=C1C4=CC=C(O)C=C4
Molecular Weight (g/mol) 386.44
Formula Weight 386.44
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.