missing translation for 'onlineSavingsMsg'
Learn More

Propyl Parahydroxybenzoate 0.01 mg/ml in Methanol, LoGiCal

Product Code. 30458828
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458828 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458828 Supplier LoGiCal Supplier No. LGCAMP0432.0004

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Propyl Parahydroxybenzoate 0.01 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propyl Parahydroxybenzoate
CAS 94-13-3
Analyte or Component Names Propyl Parahydroxybenzoate
Shipping Condition Room Temperature
Molecular Formula C10 H12 O3
Concentration 0.01 mg/mL
Certifications/Compliance ISO 17034
Synonym Propyl 4-Hydroxybenzoate, Propyl Parahydroxybenzoate, Ethyl Parahydroxybenzoate Imp. C (EP), Butyl Parahydroxybenzoate Imp. D (EP), Methyl Parahydroxybenzoate Imp. C (EP), Nipasol M, SKF-4392, Propyl Paraben, Butyl Parahydroxybenzoate Impurity D, Ethyl Parahydroxybenzoate Impurity C, Methyl Parahydroxybenzoate Impurity C
InChI Formula InChI=1 S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11 H,2,7H2,1H3
SMILES CCCOC(=O)c1ccc(O)cc1
IUPAC Name propyl 4-hydroxybenzoate
Solvent or Matrix Methanol
Industry Type Food Analysis
Molecular Weight (g/mol) 180.2
Quantity 1 mL
Formula Weight 180.0786 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.