missing translation for 'onlineSavingsMsg'
Learn More

Propyl isothiocyanate, 97%, Thermo Scientific™

Product Code. 10317504
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10317504 10 g Amber Glass Bottle 10g
10634025 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10317504 Supplier Thermo Scientific Maybridge Supplier No. TL01934EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 628-30-8
Molecular Formula C4H7NS
Molecular Weight (g/mol) 101.167
MDL Number MFCD00004823
InChI Key KKASGUHLXWAKEZ-UHFFFAOYSA-N
Synonym propyl isothiocyanate, propylisothiocyanate, propane, 1-isothiocyanato, n-propyl isothiocyanate, isothiocyanic acid propyl ester, isothiocyanic acid, propyl ester, isothiocyanic acid n-propyl ester, 1-propyl isothiocyanate, propanisothiocyanate, 1-isothiocyanatopropan
PubChem CID 69403
IUPAC Name 1-isothiocyanatopropane
SMILES CCCN=C=S

Specifications

CAS Min % 97.0
Density 0.9780g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula CH3CH2CH2NCS
Quantity 10 g
Formula Weight 101.17
Percent Purity 97%
Chemical Name or Material Propyl isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.