missing translation for 'onlineSavingsMsg'
Learn More

Propyl disulfide, 99%, Thermo Scientific™

Produktkode 15780839 Shop alle Thermo Scientific Chemicals produkter
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode
15780839 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 15780839 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 232340050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolitik

Tekniske data

CAS 629-19-6
Density 0.9500g/mL
Boiling Point 195.0°C to 196.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H14S2
Linear Formula [CH3(CH2)2S]2
Refractive Index 1.4940 to 1.5000
MDL Number MFCD00009378
Quantity 5g
Specific Gravity 0.95
Synonym dipropyl disulfide, propyl disulfide, disulfide, dipropyl, n-propyl disulfide, di-n-propyl disulfide, 4,5-dithiaoctane, dipropyl disulphide, 1-propyldisulfanyl propane, propyldithiopropane, 1,1'-dithiodipropane
Solubility Information Solubility in water: insoluble.
InChI Key ALVPFGSHPUPROW-UHFFFAOYSA-N
SMILES CCCSSCCC
IUPAC Name 1-(propyldisulfanyl)propane
Molecular Weight (g/mol) 150.30
PubChem CID 12377
ChEBI CHEBI:45758
Formula Weight 150.24
Percent Purity 99%
Chemical Name or Material Propyl disulfide, 99%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.