missing translation for 'onlineSavingsMsg'
Learn More

Propyl chloroformate, 98%

Product Code. 10692835 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10692835 25 g 25g
10471294 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10692835 Supplier Thermo Scientific Acros Supplier No. 245290250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 109-61-5 | C4H7ClO2 | 122.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-61-5
Molecular Weight (g/mol) 122.55
InChI Key QQKDTTWZXHEGAQ-UHFFFAOYSA-N
Synonym propyl chlorocarbonate, n-propyl chloroformate, formic acid, chloro-, propyl ester, carbonochloridic acid, propyl ester, propylchloroformate, propyl chloroformate, unii-a5y2t003ju, chloroformicacidpropylester, n-propylchloroformate, chloroformic acid propyl ester
PubChem CID 7998
IUPAC Name propyl carbonochloridate

Specifications

Melting Point -70.0°C
Density 1.0900 L532g/mL
Boiling Point 106.0°C
Flash Point 16°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4H7ClO2
Linear Formula CH3(CH2)2OCOCl
Refractive Index 1.4030 to 1.4050
Quantity 25 g
Merck Index 15,7961
Specific Gravity 1.09
Solubility Information Solubility in water: reacts
SMILES CCCOC(Cl)=O
Viscosity 0.70 mPa.s (20°C)
Formula Weight 122.55
Percent Purity 98%
Chemical Name or Material Propyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.