missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propyl Acetate, ≥99.5%, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Melting Point | -95°C |
Color | Colorless |
Density | 0.888g/mL at 25°C |
Boiling Point | 102°C |
Flash Point | 14°C (57.2°F) |
Ignition Point | 842°F |
Packaging | Glass Bottle |
Linear Formula | CH3COOCH2CH2CH3 |
Quantity | 1 L |
UN Number | UN1276 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'