missing translation for 'onlineSavingsMsg'
Learn More

Propyl 4-Hydroxybenzoate(Propyl Paraben), TRC

Product Code. 30004861
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004861 25 g 25g
30005286 10 g 10g
17930085 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004861 Supplier Toronto Research Chemicals Supplier No. TRCP83828525G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-13-3
Molecular Formula C10 H12 O3
Molecular Weight (g/mol) 180.2
Synonym Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Ethyl Parahydroxybenzoate Imp. C (EP), Butyl Parahydroxybenzoate Imp. D (EP), Methyl Parahydroxybenzoate Imp. C (EP), Benzoic acid, 4-hydroxy-, propyl ester, Benzoic acid, p-hydroxy-, propyl ester (6CI,8CI), 4-Hydroxybenzoic acid propyl ester, Aseptoform P, Bonomold OP, Chemocide PK, Mekkings P, NSC 23515, NSC 8511, Nipagin P, Nipasol, Nipasol M, Nipasol P, Nipazol, Paraben P, Paratexin P, Paseptol, Preserval P, Propagin, Propyl paraben, Propyl Butex, Propyl Chemosept, Propyl p-hydroxybenzoate, Propylparaben, Propylparasept, Pulvis conservans, Solbrol P, Tegasept P, Tegosept P, n-Propyl 4-hydroxybenzoate, n-Propylparaben, p-Hydroxybenzoic acid propyl ester, p-Hydroxybenzoic propyl ester, propyl p-oxybenzoate
IUPAC Name propyl 4-hydroxybenzoate
SMILES CCCOC(=O)c1ccc(O)cc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C10H12O3/c1-2-7-13-10(12)8-3-5-9(11)6-4-8/h3-6,11H,2,7H2,1H3
Formula Weight 180.0786
Percent Purity >95%
Chemical Name or Material Propyl Parahydroxybenzoate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.