missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proprotein Convertase 9/PCSK9 Rabbit anti-Human, Clone: A1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89576-50UL
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for ELISA.
Specifications
| Proprotein Convertase 9/PCSK9 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| A1 | |
| ELISA 1:1000-1:2000 | |
| EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| Recombinant Human Proprotein Convertase 9/PCSK9 protein | |
| 50 μg | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 255738 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction