missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proprotein Convertase 9/PCSK9 Antibody (Schering patent anti-PCSK9) [Janelia Fluor™ 525], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28202JF525
This item is not returnable.
View return policy
Description
Proprotein Convertase 9/PCSK9 Monoclonal antibody specifically detects Proprotein Convertase 9/PCSK9 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Proprotein Convertase 9/PCSK9 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9 | |
| PCSK9 | |
| 0.1 mL | |
| Cholesterol Metabolism, Lipid and Metabolism | |
| 255738 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Schering patent anti-PCSK9 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction