missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Proprotein Convertase 9/PCSK9 Antibody Pair [HRP]

Product Code. 18684660 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18684660

Brand: Novus Biologicals™ NBP2796185Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Proprotein Convertase 9/PCSK9.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 23.44 pg/mL
Assay Sensitivity 23.44-1500 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Rat Proprotein Convertase 9/PCSK9.
Target Species Rat
For Use With (Application) Sandwich ELISA
Gene Alias EC 3.4.21, EC 3.4.21.111, FH3neural apoptosis regulated convertase 1, HCHOLA3, hypercholesterolemia, autosomal dominant 3, LDLCQ1, NARC-1convertase subtilisin/kexin type 9 preproprotein, NARC1EC 3.4.21.-, Neural apoptosis-regulated convertase 1, PC9, Proprotein convertase 9, proprotein convertase subtilisin/kexin type 9, Subtilisin/kexin-like protease PC9
Gene ID (Entrez) 255738
Gene Symbol PCSK9
Host Species Rabbit/Mouse
Kit Contents
  • Standard: Each vial contains 23 ng of recombinant Rat Proprotein Convertase 9/PCSK9.
  • Capture Antibody: 0.5 mg/mL of mouse anti-Rat Proprotein Convertase 9/PCSK9 monoclonal antibody (in PBS, pH 7.4).
  • Detection Antibody: 0.2 mg/mL of rabbit anti-Proprotein Convertase 9/PCSK9 monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4, store at 4°C).
  • Quantity 5 Plates
    Regulatory Status RUO
    Research Discipline Cholesterol Metabolism, Lipid and Metabolism
    Storage Requirements Storage is content dependent.
    Target Proprotein Convertase 9/PCSK9
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in PBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 ug/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20°C to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1500 pg/mL is recommended.
    Show More Show Less
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.