missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proprotein Convertase 2/PCSK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48942
This item is not returnable.
View return policy
Description
Proprotein Convertase 2/PCSK2 Polyclonal antibody specifically detects Proprotein Convertase 2/PCSK2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Proprotein Convertase 2/PCSK2 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 3.4.21, EC 3.4.21.94, KEX2-like endoprotease 2, NEC 2, NEC2SPC2, neuroendocrine convertase 2, PC2NEC-2, Prohormone convertase 2, Proprotein convertase 2, proprotein convertase subtilisin/kexin type 2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: FTNHFLVELHKGGEDKARQVAAEHGFGVRKLPFAEGLYHFYHNGLAKAKRRRSLHHKQQLERDPRVKMALQQEGFDRKKRGYRDINEIDINMNDPLFTKQWYLINTGQADGTPGLDLNVAEAWELGYTGKGVTIGIMDDGIDYLHP | |
| 0.1 mL | |
| Chromatin Research, Golgi Apparatus Markers, Neuroscience | |
| 5126 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction