missing translation for 'onlineSavingsMsg'
Learn More

Propranolol hydrochloride, MedChemExpress

Product Code. p-200101482
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16452239

Brand: Medchem Express HYB0573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Propranolol hydrochloride is a nonselective β-adrenergic receptor (βAR) antagonist, has high affinity for the β1AR and β2AR with Ki values of 1.8 nM and 0.8 nM, respectively.

Propranolol hydrochloride inhibits [3H]-DHA binding to rat brain membrane preparation with an IC50 of 12 nM. Propranolol hydrochloride is used for study of hypertension, pheochromocytoma, myocardial infarction, cardiac arrhythmias, angina pectoris, and hypertrophic cardiomyopathy.

Specifications

Chemical Name or Material Propranolol hydrochloride
CAS 318-98-9
Color White
Purity Grade Notes Research
Molecular Formula C16H22ClNO2
Quantity 1 g
Solubility Information DMSO : ≥ 150 mg/mL (507.10 mM) ∣H2O : 33.33 mg/mL (112.68 mM; Need ultrasonic)
SMILES OC(CNC(C)C)COC1=CC=CC2=CC=CC=C12.Cl
Molecular Weight (g/mol) 295.8
Formula Weight 295.8
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.