missing translation for 'onlineSavingsMsg'
Learn More

Propoxycaine Hydrochloride, TRC

Product Code. 30424751
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424751 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424751 Supplier Toronto Research Chemicals Supplier No. TRCP831300100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propoxycaine Hydrochloride
CAS 550-83-4
Molecular Formula C16 H26 N2 O3 . Cl H
Quantity 100 mg
Synonym Benzoic acid','4-amino-2-propoxy-','2-(diethylamino)ethyl ester','hydrochloride (1:1), Benzoic acid','4-amino-2-propoxy-','2-(diethylamino)ethyl ester','monohydrochloride (8CI,9CI), 2-Diethylaminoethyl 4-amino-2-propoxybenzoate hydrochloride, 2'-Diethylaminoethyl 2-propoxy-4-aminobenzoate hydrochloride, 4-Amino-2-propoxybenzoic acid 2-diethylaminoethyl ester hydrochloride, Blockain hydrochloride, Blockaine hydrochloride, Pravocaine hydrochloride, Propoxycaine hydrochloride, Ravocaine hydrochloride, Rovocaine hydrochloride
InChI Formula InChI=1S/C16H26N2O3.ClH/c1-4-10-20-15-12-13(17)7-8-14(15)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H
SMILES Cl.CCCOc1cc(N)ccc1C(=O)OCCN(CC)CC
IUPAC Name 2-(diethylamino)ethyl 4-amino-2-propoxybenzoate;hydrochloride
Molecular Weight (g/mol) 330.85
Formula Weight 330.171
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.