missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propoxycaine Hydrochloride, Mikromol™
Description
Propoxycaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Propoxycaine Hydrochloride |
| CAS | 550-83-4 |
| Analyte or Component Names | Propoxycaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H26 N2 O3 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Benzoic acid, 4-amino-2-propoxy-, 2-(diethylamino)ethyl ester, hydrochloride (1:1), Benzoic acid, 4-amino-2-propoxy-, 2-(diethylamino)ethyl ester, monohydrochloride (8CI,9CI), 2-Diethylaminoethyl 4-amino-2-propoxybenzoate hydrochloride, 2'-Diethylaminoethyl 2-propoxy-4-aminobenzoate hydrochloride, 4-Amino-2-propoxybenzoic acid 2-diethylaminoethyl ester hydrochloride, Blockain hydrochloride, Blockaine hydrochloride, Pravocaine hydrochloride, Propoxycaine hydrochloride, Ravocaine hydrochloride, Rovocaine hydrochloride |
| InChI Formula | InChI=1S/C16H26N2O3.ClH/c1-4-10-20-15-12-13(17)7-8-14(15)16(19)21-11-9-18(5-2)6-3;/h7-8,12H,4-6,9-11,17H2,1-3H3;1H |
| SMILES | Cl.CCCOc1cc(N)ccc1C(=O)OCCN(CC)CC |
| IUPAC Name | 2-(diethylamino)ethyl 4-amino-2-propoxybenzoate;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?