missing translation for 'onlineSavingsMsg'
Learn More

Propoxycaine hydrochloride, MedChemExpress

Product Code. 16424577
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16424577 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16424577 Supplier Medchem Express Supplier No. HYB1243

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Propoxycaine hydrochloride inhibits voltage-gated sodium channels, and thereby inhibits the ionic flux required for the initiation and conduction of impulses. Propoxycaine hydrochloride application can lead to a loss of sensation.

Propoxycaine hydrochloride inhibits voltage-gated sodium channels, and thereby inhibits the ionic flux required for the initiation and conduction of impulses. Propoxycaine hydrochloride application can lead to a loss of sensation.

Specifications

Chemical Name or Material Propoxycaine hydrochloride
CAS 550-83-4
Color Light Nude
Purity Grade Notes Research
Molecular Formula C16H27ClN2O3
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (302.25 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (151.13 mM; Need ultrasonic)
SMILES O=C(OCCN(CC)CC)C1=CC=C(N)C=C1OCCC.[H]Cl
Molecular Weight (g/mol) 330.85
Formula Weight 330.85
Percent Purity 98.06%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.