missing translation for 'onlineSavingsMsg'
Learn More

Propofol, TRC

Product Code. 17914605
Change view
Click to view available options
Quantity:
25 mg
250 mg
5 g
Unit Size:
250mg
25mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17914605 250 mg 250mg
17981015 5 g 5g
17992025 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17914605 Supplier Toronto Research Chemicals Supplier No. TRCP829750250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2078-54-8
Molecular Formula C12 H18 O
Molecular Weight (g/mol) 178.27
Synonym 2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, 2,6-Diisopropylphenol, Ampofol, Diprifusor, Diprivan, Diprivan 10, Diprofol, Disoprivan, Disoprofol, ICI 35868, Ivofol, NSC 5105, PD 18215, Propofol, Propofol-lipuro, Rapinovet, Recofol
IUPAC Name 2,6-di(propan-2-yl)phenol
SMILES CC(C)c1cccc(C(C)C)c1O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13H,1-4H3
Formula Weight 178.1358
Percent Purity >95%
Chemical Name or Material Propofol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.