missing translation for 'onlineSavingsMsg'
Learn More

Propofol 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30457984
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457984 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457984 Supplier LoGiCal Supplier No. LGCAMP0461.0001
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Propofol 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propofol
CAS 2078-54-8
Analyte or Component Names Propofol
Shipping Condition Room Temperature
Molecular Formula C12 H18 O
Certifications/Compliance ISO 17034
Synonym 2,6-Bis(1-methylethyl)phenol, 2,6-Bis(isopropyl)phenol, 2,6-Diisopropylphenol, Ampofol, Diprifusor, Diprivan, Diprivan 10, Diprofol, Disoprivan, Disoprofol, ICI 35868, Ivofol, NSC 5105, PD 18215, Propofol, Propofol-lipuro, Rapinovet, Recofol
InChI Formula InChI=1 S/C12H18O/c1-8(2)10-6-5-7-11(9(3)4)12(10)13/h5-9,13 H,1-4H3
SMILES CC(C)c1cccc(C(C)C)c1O
IUPAC Name 2,6-di(propan-2-yl)phenol
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 178.27
Quantity 1 mL
Formula Weight 178.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.