missing translation for 'onlineSavingsMsg'
Learn More

Propionyl Glycine, TRC

Product Code. 30134740
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134740 5 mg 5mg
30136635 1 mg 1mg
30010175 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134740 Supplier Toronto Research Chemicals Supplier No. TRCP7885005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21709-90-0
Molecular Formula C5 H9 N O3
Molecular Weight (g/mol) 131.13
Synonym Glycine, N-(1-oxopropyl)-, Glycine, N-propionyl- (8CI), N-(1-Oxopropyl)glycine, (Propionylamino)acetic acid, N-Propionylglycine, NSC 158539, Propionylglycine
IUPAC Name 2-(propanoylamino)acetic acid
SMILES CCC(=O)NCC(=O)O

Specifications

InChI Formula InChI=1S/C5H9NO3/c1-2-4(7)6-3-5(8)9/h2-3H2,1H3,(H,6,7)(H,8,9)
Quantity 5 mg
Formula Weight 131.0582
Shipping Condition Room Temperature
Chemical Name or Material Propionyl Glycine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.