missing translation for 'onlineSavingsMsg'
Learn More

Propionitrile, TRC

Product Code. 30423366
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423366 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423366 Supplier Toronto Research Chemicals Supplier No. TRCP7963902.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionitrile
CAS 107-12-0
Molecular Formula C3 H5 N
Quantity 2.5 g
Synonym Propanenitrile, Propionitrile (8CI), 1-Methylacetonitrile, Cyanoethane, Ether cyanatus, Ethyl cyanide, Hydrocyanic ether, NSC 7966, Propionic nitrile, Propiononitrile, Propylnitrile, n-Propanenitrile
InChI Formula InChI=1S/C3H5N/c1-2-3-4/h2H2,1H3
SMILES CCC#N
IUPAC Name propanenitrile
Molecular Weight (g/mol) 55.08
Formula Weight 55.0422
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.