missing translation for 'onlineSavingsMsg'
Learn More

Propionic Anhydride, TRC

Product Code. 30410282
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30410282 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30410282 Supplier Toronto Research Chemicals Supplier No. TRCP7863805G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionic Anhydride
CAS 123-62-6
Molecular Formula C6 H10 O3
Quantity 5 g
Synonym Propanoic acid','1,1'-anhydride, Propanoic acid','anhydride (9CI), Propionic anhydride (6CI,8CI), Methylacetic anhydride, Propanoic anhydride, Propanoyl propanoate, Propionic acid anhydride, Propionyl anhydride, Propionyl oxide
InChI Formula InChI=1S/C6H10O3/c1-3-5(7)9-6(8)4-2/h3-4H2,1-2H3
SMILES CCC(=O)OC(=O)CC
IUPAC Name propanoyl propanoate
Molecular Weight (g/mol) 130.1418
Formula Weight 130.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.