missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid Methyl Ester, TRC

Product Code. 30426552
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426552 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30426552 Supplier Toronto Research Chemicals Supplier No. TRCP78637010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propionic acid methyl ester
CAS 554-12-1
Molecular Formula C4 H8 O2
Quantity 10 g
Synonym Propanoic acid','methyl ester, Propionic acid','methyl ester (8CI), Propanoic acid methyl ester, Propionic acid methyl ester, Methyl propanoate, Methyl propionate, Methyl propylate, NSC 9375
InChI Formula InChI=1S/C4H8O2/c1-3-4(5)6-2/h3H2,1-2H3
SMILES CCC(=O)OC
IUPAC Name methyl propanoate
Molecular Weight (g/mol) 88.11
Formula Weight 88.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.