missing translation for 'onlineSavingsMsg'
Learn More

Propionic Acid, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7073462 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10345860

Brand: Fisher Chemical P/7560/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 79-09-4
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.079
MDL Number 2756
InChI Key XBDQKXXYIPTUBI-UHFFFAOYSA-N
Synonym propionic acid, ethylformic acid, methylacetic acid, carboxyethane, ethanecarboxylic acid, pseudoacetic acid, metacetonic acid, monoprop, luprosil, prozoin
PubChem CID 1032
ChEBI CHEBI:30768
IUPAC Name propanoic acid
SMILES CCC(=O)O

Specifications

Melting Point -22°C
pH 2.5
Boiling Point 141°C
Packaging HDPE plastic bottle
Quantity 500 mL
Vapor Density 2.56
Formula Weight 74.08g/mol
Vapor Pressure 5mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propionic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.