missing translation for 'onlineSavingsMsg'
Learn More

Propionic acid-ethyl ester, Dr. Ehrenstorfer

Product Code. 30315494
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30315494 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315494 Supplier Dr. Ehrenstorfer Supplier No. DRECA16493500100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material Propanoic acid ethyl ester
CAS 105-37-3
Analyte or Component Names Propanoic acid ethyl ester
Shipping Condition +20°C
Molecular Formula C5 H10 O2
Certifications/Compliance ISO 17034
Synonym Propionic acid-ethyl ester
InChI Formula InChI=1S/C5H10O2/c1-3-5(6)7-4-2/h3-4H2,1-2H3
SMILES CCOC(=O)CC
IUPAC Name ethyl propanoate
Molecular Weight (g/mol) 102.13
Quantity 100 mg
Formula Weight 102.0681 g/mol
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.