missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, 97%

Product Code. 10498670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
25mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10498670 1 L Glass bottle 1 L
10553681 25 mL Glass bottle 25mL
10408680 2.5 L Glass bottle 2.5 L
11994771 10 L Plastic drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10498670 Supplier Thermo Scientific Acros Supplier No. 131510010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-38-6 | C3H6O | 58.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
InChI Key NBBJYMSMWIIQGU-UHFFFAOYSA-N
Synonym propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
PubChem CID 527
ChEBI CHEBI:17153
IUPAC Name propanal
SMILES CCC=O

Specifications

Melting Point -81.0°C
Color Colorless
Density 0.7980g/mL
Boiling Point 48.0°C
Flash Point −40°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CHO
Refractive Index 1.3610 to 1.3650
Quantity 1 L
Beilstein 01,629
Merck Index 15,7937
Specific Gravity 0.798
Solubility Information Solubility in water: 540g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.6 mPa.s (20°C)
Formula Weight 58.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Propionaldehyde
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propionaldehyde
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.