missing translation for 'onlineSavingsMsg'
Learn More

Propionaldehyde, 97%, Honeywell™

Product Code. 15675800 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15675800 1 L Glass bottle 1 L
15685800 250 mL Glass bottle 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15675800 Supplier Honeywell Research Chemicals Supplier No. 5381241L

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 97%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-38-6
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00007020
InChI Key NBBJYMSMWIIQGU-UHFFFAOYSA-N
Synonym propionaldehyde, propanaldehyde, propaldehyde, propional, propionic aldehyde, n-propanal, methylacetaldehyde, propylaldehyde, propylic aldehyde, aldehyde propionique
PubChem CID 527
ChEBI CHEBI:17153
IUPAC Name propanal
SMILES CCC=O

Specifications

Melting Point -81°C
Color Colorless
Density 0.805g/mL at 25°C
Boiling Point 46°C to 50°C
Ignition Point 404°C
Packaging Glass bottle
Linear Formula CH3CH2CHO
Quantity 1 L
UN Number UN1275
Beilstein 506010
Vapor Density 2 (vs air)
Formula Weight 58.08g/mol
Vapor Pressure 18.77mmHg (55°C), 4.89mmHg (20°C)
Percent Purity 0.97
Chemical Name or Material Propionaldehyde
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.