missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid, TRC

Product Code. 30428662
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30428662 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30428662 Supplier Toronto Research Chemicals Supplier No. TRCP8278605G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propiolic Acid
CAS 471-25-0
Molecular Formula C3H2O2
Quantity 5 g
Synonym 2-Propyne-1-carboxylic Acid, Acetylenecarboxylic Acid, Acetylenemonocarboxylic Acid, Carboxyacetylene, NSC 16152, Propargylic Acid, Propynoic Acid
InChI Formula InChI=1S/C3H2O2/c1-2-3(4)5/h1H,(H,4,5)
SMILES OC(=O)C#C
IUPAC Name prop-2-ynoic acid
Molecular Weight (g/mol) 70.05
Formula Weight 70.0055
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.