missing translation for 'onlineSavingsMsg'
Learn More

Propiolic Acid, 98%

Product Code. 10413255 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
10mL
25mL
This item is not returnable. View return policy

Product Code. 10413255

Brand: Thermo Scientific Acros 131501000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-25-0 | C3H2O2 | 70.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 16.0°C
Color Colorless to Brown
Density 1.1300g/mL
Boiling Point 153.0°C to 157.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HC≡CCO2H
Refractive Index 1.4315 to 1.4335
Quantity 100 mL
Merck Index 15, 7935
Specific Gravity 1.13
Solubility Information Solubility in water: miscible
Formula Weight 70.04
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Propiolic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.