missing translation for 'onlineSavingsMsg'
Learn More

Propiolaldehyde diethyl acetal, 97%, Thermo Scientific Chemicals

£34.10 - £316.00

Chemical Identifiers

CAS 10160-87-9
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00009237
InChI Key RGUXEWWHSQGVRZ-UHFFFAOYSA-N
PubChem CID 66285
IUPAC Name 3,3-diethoxyprop-1-yne
SMILES CCOC(OCC)C#C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332986
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10767.03
1 g
£34.10
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11342986
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10767.06
5 g
£97.50
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11352986
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L10767.14
25 g
£316.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propiolaldehyde diethyl acetal is used in preparation of 3-boronoacrolein pinacolate, a heterodyne and (E)-3-(tributylstannyl)-2-propenal.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature is 2 - 8°C. Air and moisture sensitive. Store under inert gas.
Specifications

Chemical Identifiers

10160-87-9
128.17
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
3,3-diethoxyprop-1-yne
C7H12O2
MFCD00009237
66285
CCOC(OCC)C#C

Specifications

10160-87-9
138°C to 139°C
C7H12O2
MFCD00009237
1701566
Slightly soluble in water.
CCOC(OCC)C#C
128.17
128.17
Propiolaldehyde diethyl acetal
0.894
32°C (89°F)
1.412
UN1993
3,3-diethoxy-1-propyne, propargylaldehyde diethyl acetal, 3,3-diethoxypropyne, propiolaldehyde diethyl acetal, 1-propyne, 3,3-diethoxy, 3,3-diethoxy-propyne, propynal diethyl acetal, 1,1-diethoxyprop-2-yne, 1-propyne,3,3-diethoxy, propargyl aldehyde diethylacetal
RGUXEWWHSQGVRZ-UHFFFAOYSA-N
3,3-diethoxyprop-1-yne
66285
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 233-430-4

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only