missing translation for 'onlineSavingsMsg'
Learn More

Propidium Iodide, TRC

Product Code. 30419231
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419231 25 mg -
30416661 50 mg -
30415369 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30419231 Supplier Toronto Research Chemicals Supplier No. TRCP77050025MG
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27 H34 N4 . 2 I
Molecular Weight (g/mol) 668.39
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES [I-].[I-].CC[N+](C)(CC)CCC[n+]1c(c2ccccc2)c3cc(N)ccc3c4ccc(N)cc14

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C27H33N4.2HI/c1-4-31(3,5-2)17-9-16-30-26-19-22(29)13-15-24(26)23-14-12-21(28)18-25(23)27(30)20-10-7-6-8-11-20;;/h6-8,10-15,18-19,29H,4-5,9,16-17,28H2,1-3H3;2*1H/q+1;;/p-1
Formula Weight 668.0873
Chemical Name or Material Propidium Iodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.