missing translation for 'onlineSavingsMsg'
Learn More

Properdin Antibody [PE], Novus Biologicals™

Product Code. 30737428 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30737428 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737428 Supplier Novus Biologicals Supplier No. AF8216P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Properdin Polyclonal antibody specifically detects Properdin in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Properdin
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias BFD, Complement factor P, complement factor properdin, PFCcomplement, PFD, PROPERDIN
Host Species Sheep
Immunogen Human embryonic kidney cell line HEK293-derived human Properdin, Asp28-Leu469, Accession # P27918
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 5199
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.