missing translation for 'onlineSavingsMsg'
Learn More

Propargyloxytrimethylsilane, 97%, Thermo Scientific™

Product Code. 10229643 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10229643 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10229643 Supplier Thermo Scientific Acros Supplier No. 313900025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 5582-62-7
Color Yellow to Brown
Density 0.8300g/mL
Boiling Point 110.0°C
Flash Point 8°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C6H12OSi
Linear Formula HC≡CCH2OSi(CH3)3
Refractive Index 1.4070 to 1.4090
MDL Number MFCD00042844
Quantity 2.5 g
Specific Gravity 0.83
Synonym propargyloxytrimethylsilane, propargyloxy trimethylsilane, o-trimethylsilylpropargyl alcohol, silane, trimethyl 2-propynyloxy, trimethyl prop-2-yn-1-yloxy silane, 3-trimethylsiloxy-1-propyne, trimethyl 2-propynyloxy silane, silane, trimethyl 2-propyn-1-yloxy, trimethyl prop-2-ynyloxy silane, 3-trimethylsiloxypropyne
Solubility Information Solubility in water: insoluble.
InChI Key ZZRPJWCNCLSOLR-UHFFFAOYSA-N
SMILES C[Si](C)(C)OCC#C
IUPAC Name trimethyl(prop-2-ynoxy)silane
Molecular Weight (g/mol) 128.25
PubChem CID 79696
Formula Weight 128.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Propargyloxytrimethylsilane, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.