missing translation for 'onlineSavingsMsg'
Learn More

(Propargyloxy)trimethylsilane, 97%

Product Code. 11347806
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347806 5 g 5g
11357806 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347806 Supplier Thermo Scientific Alfa Aesar Supplier No. L12936.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5582-62-7 | C6H12OSi | 128.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5582-62-7
Molecular Formula C6H12OSi
Molecular Weight (g/mol) 128.246
MDL Number MFCD00042844
InChI Key ZZRPJWCNCLSOLR-UHFFFAOYSA-N
Synonym propargyloxytrimethylsilane, propargyloxy trimethylsilane, o-trimethylsilylpropargyl alcohol, silane, trimethyl 2-propynyloxy, trimethyl prop-2-yn-1-yloxy silane, 3-trimethylsiloxy-1-propyne, trimethyl 2-propynyloxy silane, silane, trimethyl 2-propyn-1-yloxy, trimethyl prop-2-ynyloxy silane, 3-trimethylsiloxypropyne
PubChem CID 79696
IUPAC Name trimethyl(prop-2-ynoxy)silane
SMILES C[Si](C)(C)OCC#C

Specifications

Boiling Point 110°C to 111°C
Quantity 5 g
Flash Point 8°C (46°F)
Refractive Index 1.408
Sensitivity Moisture sensitive
Density 0.833
Linear Formula (CH3)3SiOCH2C≡CH
UN Number UN1993
Beilstein 2036793
Formula Weight 128.25
Percent Purity 97%
Chemical Name or Material (Propargyloxy)trimethylsilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.