missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine hydrochloride, 95%

Product Code. 11380408
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11380408 1 g 1g
11390408 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11380408 Supplier Thermo Scientific Alfa Aesar Supplier No. 043775.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15430-52-1 | C3H6ClN | 91.54 g/mol

Propargylamine hydrochloride is a useful intermediates in the preparation of drugs. It is used in pharmaceutical, industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15430-52-1
Molecular Formula C3H6ClN
Molecular Weight (g/mol) 91.54
MDL Number MFCD00012907
InChI Key IKXNIQJDNKPPCH-UHFFFAOYSA-N
Synonym propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
PubChem CID 11205720
IUPAC Name prop-2-yn-1-amine;hydrochloride
SMILES [H+].[Cl-].NCC#C

Specifications

Melting Point 179°C to 182°C
Quantity 1 g
UN Number UN2811
Sensitivity Hygroscopic
Solubility Information Soluble in water (partly),methanol,and ethanol.
Formula Weight 91.54
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Propargylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.