missing translation for 'onlineSavingsMsg'
Learn More

Propargylamine hydrochloride, 95%

Product Code. 10336780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10336780 1 g Glass bottle 1g
10351624 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10336780 Supplier Thermo Scientific Acros Supplier No. 131470010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15430-52-1 | C3H6ClN | 91.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15430-52-1
Molecular Formula C3H6ClN
Molecular Weight (g/mol) 91.54
MDL Number MFCD00012907
InChI Key IKXNIQJDNKPPCH-UHFFFAOYSA-N
Synonym propargylamine hydrochloride, prop-2-yn-1-amine hydrochloride, 2-propynylamine hydrochloride, propargylamine hcl, 2-propynylamine hcl, 3-amino-1-propyne hydrochloride, prop-2-ynylamine hydrochloride, 2-propyn-1-amine, hydrochloride, pubchem18845, acmc-1bvqh
PubChem CID 11205720
IUPAC Name prop-2-yn-1-amine;hydrochloride
SMILES [H+].[Cl-].NCC#C

Specifications

Melting Point 175°C to 182°C
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Linear Formula HC∽CCH2NH2·HCl
Quantity 1 g
Beilstein 04, 228
Formula Weight 91.54
Percent Purity 95%
Chemical Name or Material Propargylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.