Learn More
Propargylamine, 98%, Thermo Scientific Chemicals
£62.90 - £207.00
Chemical Identifiers
CAS | 2450-71-7 |
---|---|
Molecular Formula | C3H5N |
Molecular Weight (g/mol) | 55.08 |
MDL Number | MFCD00008198 |
InChI Key | JKANAVGODYYCQF-UHFFFAOYSA-N |
Synonym | propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine |
PubChem CID | 239041 |
IUPAC Name | prop-2-yn-1-amine |
SMILES | C#CCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15470378
|
Thermo Scientific Alfa Aesar
H53495.06 |
5 g |
£62.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480378
|
Thermo Scientific Alfa Aesar
H53495.14 |
25 g |
£207.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPropargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.
Solubility
Miscible with water, chloroform and ethyl acetate.
Notes
Hygroscopic. Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
Chemical Identifiers
2450-71-7 | |
55.08 | |
JKANAVGODYYCQF-UHFFFAOYSA-N | |
239041 | |
C#CCN |
C3H5N | |
MFCD00008198 | |
propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine | |
prop-2-yn-1-amine |
Specifications
2450-71-7 | |
83°C | |
C3H5N | |
1.449 | |
UN3286 | |
Air Sensitive | |
Miscible with water,chloroform and ethyl acetate. | |
C#CCN | |
55.08 | |
55.08 | |
Propargylamine |
0.86 | |
6°C (43°F) | |
HC≡CCH2NH2 | |
MFCD00008198 | |
773681 | |
propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine | |
JKANAVGODYYCQF-UHFFFAOYSA-N | |
prop-2-yn-1-amine | |
239041 | |
98% |
Safety and Handling
GHS H Statement
H225-H310-H314-H318-H302
Highly flammable liquid and vapor.
Fatal in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302-H311-H314
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUID, TOXIC, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 219-513-8
missing translation for 'rtecsNumber' : UK5250000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only