missing translation for 'onlineSavingsMsg'
Learn More

Propargyl propionate, 98%, Thermo Scientific™

Product Code. 11317386
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317386 10 g 10g
11327386 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317386 Supplier Thermo Scientific Alfa Aesar Supplier No. L09371.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1932-92-9
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.128
MDL Number MFCD00039917
InChI Key BGRYSGVIVVUJHH-UHFFFAOYSA-N
Synonym propargyl propionate, 2-propyn-1-ol, propanoate, 2-propynyl propionate, 2-propyn-1-ol, propionate, propargyl proprionate, 2-propyn-1-ol, 1-propanoate, prop-2-yn-1-yl propanoate, propanoic acid, 2-propynyl ester, propargylpropionate, acmc-20ajth
PubChem CID 74735
IUPAC Name prop-2-ynyl propanoate
SMILES CCC(=O)OCC#C

Specifications

Density 0.973
Boiling Point 137°C to 139°C
Flash Point 40°C (104°F)
Refractive Index 1.423
Quantity 10 g
UN Number UN3272
Beilstein 1902510
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material Propargyl propionate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.